Docosanal 98%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

H33924.MD H33924.03
H33924.MDEA 1150 DKK
H33924.MD H33924.03
Docosanal 98%
Docosanal
Formel: C₂₂H₄₄O
Molekylvægt: 324.593 g/mol
Smeltepunkt: 57…60 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD07780595
CAS-nummer: 57402-36-5

Bestil nu

Specifikationer Testresultater

Assay Minimum 98


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR