ThermoSafe® AmbiTech® Pre-Qualified 15 to 25 °C Shippers
Leverandør: Sonoco Thermosafe
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
ATG22 ATG65 ATR28
SCATATG22EA 13580 DKK
SCATATG22 SCATATG65 SCATATR28
ThermoSafe® AmbiTech® Pre-Qualified 15 to 25 °C Shippers
Transportbeholdere
AmbiTech® pre-qualified shippers use an inorganic salt-based PCM to provide exceptional temperature control.
  • Pre-qualified 15 to 25 °C shipper
  • Tested to several ambient profiles to speed up implementation.
  • Universal packout simplifies operations; no need to manage the switch from summer to winter and back again
  • Integrated handles for easy movement
AmbiTech® controlled room temperature systems use PCM to deliver superior temperature control for temperature-sensitive pharmaceuticals, even in the harshest of climates. These systems incorporate AmbiSorb®, a custom inorganic salt PCM which is both economic and non-hazardous. They provide excellent stability in extreme ambient conditions over extended durations. Simple and quick assembly with minimal components saves time and money.