Keratinocyte Growth Medium
Leverandør: PROVITRO
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2000501 2030501
PRTV2000501EA 1250 DKK
PRTV2000501 PRTV2030501
Keratinocyte Growth Medium
Celledyrkningsmedier
Keratinocyte Growth Medium is a sterile liquid serum-free culture medium designed for cultivating human keratinocytes (HKER). Delivered as a basal medium, it is suitable for culturing Provitro HKER upon the addition of the supplement mix.
  • Formulation is optimised for the initial seeding of 6000 cells/cm², supporting growth up to confluence (approximately 75%)
  • Feeder layers, matrix substrates, or additional substances are not necessary for effective cell cultivation
The Keratinocyte Growth Medium undergoes comprehensive testing after each production cycle. All components undergo rigorous testing in a stringent biological assay. Each batch is specifically examined for the proliferating characteristics of human keratinocytes (HKER). The cells cultured in the keratinocyte growth medium are thoroughly assessed for their morphology, adherence rate, colony-forming efficiency, and population doubling time.
Storge: Store the supplemented Smooth muscle cell growth medium in a dark environment at temperatures ranging from 4 to 8 °C for a maximum duration of one month. Avoid heating the medium beyond 37 °C and refrain from using uncontrolled heat sources, such as microwave appliances. When utilising a portion of the medium, extract the required amount from the bottle and warm it appropriately. Never freeze the medium, as this could result in elevated salt concentrations due to the separation of pure water during freezing, leading to irreversible damage.