Indium(III)chlorid, anhydrous ≥99.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Indium Trichloride Chloride, Indium trichloride

I0778-25G I0778-5G
TCIAI0778-25GEA 1840 DKK
TCIAI0778-25G TCIAI0778-5G
Indium(III)chlorid, anhydrous ≥99.0% (by titrimetric analysis)
Indium(III) chloride

CAS Number: 10025-82-8
MDL Number: MFCD00011058
Molecular Formula: Cl3In
Molecular Weight: 221.17
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 586

Formel: Cl₃In
Molekylvægt: 221.18 g/mol
Kogepunkt: 800
Smeltepunkt: 586 °C
MDL Number: MFCD00011058
CAS-nummer: 10025-82-8
UN nummer: 3260
ADR: 8 II
Merck Index: 13,04980

Bestil nu

Specifikationer Testresultater

Purity/Analysis Method >99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR