Empagliflozin 98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
474590010
474592500
474590010.EA
3070
DKK
474590010.
474592500.
Empagliflozin 98%
Empagliflozin
Formel:
C₂₃H₂₇ClO₇ Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD22566222 CAS-nummer: 864070-44-0 |
Specifikationer Testresultater
Appearance | white to almost white solid |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...