α-Ketoglutarsyre monokaliumsalt 97%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Potassium 4-carboxy-2-oxobutanoate

474300050 474300250 474301000
474300050.EA 428 DKK
474300050. 474300250. 474301000.
α-Ketoglutarsyre monokaliumsalt 97%
α-Ketoglutarsyre monokaliumsalt
Formel: C₅H₅KO₅
Molekylvægt: 184.19 g/mol
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00064195
CAS-nummer: 997-43-3
EINECS: 213-641-8

Bestil nu

Specifikationer Testresultater

Appearance white powder
Infrared spectrum Conforms
Titration with HClO4 96.0 to 104.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR