D,L-Anatabine 95%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 1,2,3,6-Tetrahydro-2,3'-bipyridine, (R,S)-Anatabine

473380010 473381000
473380010.EA 4890 DKK
473380010. 473381000.
D,L-Anatabine 95%
D,L-Anatabine
Formel: C₁₀H₁₂N₂
Molekylvægt: 160.22 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00078278
CAS-nummer: 2743-90-0
UN nummer: 2810
ADR: 6.1 III

Bestil nu

Specifikationer Testresultater

Appearance colorless to light yellow to orange liquid
Infrared spectrum Conforms
HPLC ≥94.0 %
Water ≤2.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR