Carrageenan
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
471060010
471062500
471060010.EA
1400
DKK
471060010.
471062500.
Carrageenan
Carrageenan
Opbevaringstemperatur:
Stue temperatur |
MDL Number:
MFCD00081480 CAS-nummer: 9000-07-1 EINECS: 232-524-2 Merck Index: 13,01878 |
Specifikationer Testresultater
Appearance | white to light beige to light brown powder |
pH | 7.5 to 10.5 (1.5% in water) |
Loss on drying | ≤12 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...