1,2,4,5-Cyclohexanetetracarboxylic dianhydride 95%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

470950050 470950250
470950050.EA 494 DKK
470950050. 470950250.
1,2,4,5-Cyclohexanetetracarboxylic dianhydride 95%
1,2,4,5-Cyclohexanetetracarboxylic dianhydride
Formel: C₁₀H₈O₆
Kogepunkt: 517,5°C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD13704912
CAS-nummer: 2754-41-8

Bestil nu

Specifikationer Testresultater

Appearance white to almost white crystalline powder
Infrared spectrum Conforms
HPLC ≥94 %
Loss on drying ≤1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR