D-Lysin 98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
469950010
469950050
469950010.EA
676
DKK
469950010.
469950050.
D-Lysin 98%
D-Lysin
Formel:
C₆H₁₄N₂O₂ Kogepunkt: 311°C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00008234 CAS-nummer: 923-27-3 EINECS: 213-091-9 |
Specifikationer Testresultater
Appearance | white to light yellow to light beige crystalline powder |
Infrared spectrum | Conforms |
Titration with HClO4 | 97.5 to 102.5 % (on dry substance) |
Water | ≤5 % |
Specific optical rotation | -12° to -16° (20°C, 589 nm) (c=1 in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...