4-Chloroquinazoline 95%

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

469400010 469400050
469400010.EA 289 DKK
469400010. 469400050.
4-Chloroquinazoline 95%
4-Chloroquinazoline
Formel: C₈H₅ClN₂
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00228682
CAS-nummer: 5190-68-1
UN nummer: 2811
ADR: 6.1,III

Bestil nu

Specifikationer Testresultater

Appearance white to light brown powder or crystals
Infrared spectrum Conforms
HPLC ≥94 %
Melting point 100°C to 105°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR