4-Iodo-3-nitrobenzamide 98%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
469161000
469165000
469161000.EA
506
DKK
469161000.
469165000.
4-Iodo-3-nitrobenzamide 98%
4-Iodo-3-nitrobenzamide
Formel:
C₇H₅IN₂O₃ Kogepunkt: 344,7°C (760mmHg) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD11110639 CAS-nummer: 160003-66-7 |
Specifikationer Testresultater
Appearance | white to beige powder |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % |
Loss on drying | ≤2 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...