Febantel

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: N-(((2-(2-MEthoxyacetamido)-4-(phenylsulphanyl)phenyl)amino)((methoxycarbonyl)imino)methyl)carbamate

468780010 468780050
468780010.EA 700 DKK
468780010. 468780050.
Febantel
Febantel
Formel: C₂₀H₂₂N₄O₆S
Opbevaringstemperatur: Fryser
MDL Number: MFCD01738527
CAS-nummer: 58306-30-2
EINECS: 261-205-0
Merck Index: 13,03973

Bestil nu

Specifikationer Testresultater

Appearance white to light yellow powder and/or crystals
Infrared spectrum Conforms
HPLC ≥97.5 %
Melting point 123°C to 129°C
Water ≤1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR