2-Methoxyestradiol

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: (8R,9S,13S,14S,17S)-2-Methoxy-13-methyl-7,8,9,11,12,13,14,15,16,17-decahydro-6H-cyclopenta[a]phenanthrene-3,17-diol

468380010 468382500
468380010.EA 4560 DKK
468380010. 468382500.
2-Methoxyestradiol
2-Methoxyestradiol
Formel: C₁₉H₂₆O₃
Molekylvægt: 302.408 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00010489
CAS-nummer: 362-07-2
UN nummer: 2811
ADR: 6.1 III

Bestil nu

Specifikationer Testresultater

Appearance white to light yellow powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Melting point 186°C to 190°C
Specific optical rotation +105° to +110° (20°C, 589 nm) (0.5% in THF)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR