4-(Bromomethyl)tetrahydropyran

Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

468280010 468280050 468280250
468280010.EA 350 DKK
468280010. 468280050. 468280250.
4-(Bromomethyl)tetrahydropyran
4-(Bromomethyl)tetrahydropyran
Formel: C₆H₁₁BrO
Molekylvægt: 179.0549 g/mol
Kogepunkt: 88…90°C (22mmHg)
Massefylde: 1.359
Opbevaringstemperatur: Fryser
MDL Number: MFCD03788561
CAS-nummer: 125552-89-8
UN nummer: 3265
ADR: 8 II

Bestil nu

Specifikationer Testresultater

Appearance almost white to light yellow liquid
Infrared spectrum Conforms
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR