Pentachlorcyclopropan 95%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
1,1,2,2,3-Pentachlorocyclopropane
416890050
416890250
416890050.EA
510
DKK
416890050.
416890250.
Pentachlorcyclopropan 95%
Pentachlorocyclopropane
Formel:
C₃HCl₅ Molekylvægt: 214.3 g/mol Kogepunkt: 55…56°C (7mmHg) Massefylde: 1.668 Flammepunkt: 100°C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00010796 CAS-nummer: 6262-51-7 |
Specifikationer Testresultater
Appearance | clear colorless to light yellow liquid |
Infrared spectrum | Conforms |
GC | ≥94 % |
Refractive index | 1.5170 to 1.5200 (20°C, 589 nm) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...