ECOSAN® Neutral Skin Wash
Leverandør: FAWECO HAUTSCHUTZ
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
13897-015 13897-004 13897-003 13897-005 13897-024 13897-033
FAWE13897-015EA 2020 DKK
FAWE13897-015 FAWE13897-004 FAWE13897-003 FAWE13897-005 FAWE13897-024 FAWE13897-033
ECOSAN® Neutral Skin Wash
Hudpleje Sæber og huden rensemidler
Neutral cleansing syndet, also suitable for the food industry.
  • Transparent skin wash
  • Colorant free
  • Soap and alkali free
  • Skin-neutral pH value
  • Perfume and fragrance free
BLUE ANGEL environmental label and EU Ecolabel for 1000 ml Neptune bottles and 2000 ml collapsible bottles.
Certificeringer: HACCP compliant.