Guldstandard, 1g/l Au i 2% saltsyre AVS TITRINORM standard til AAS
Leverandør: VWR Chemicals
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
86662.260EA
2261
DKK
86662.260
86662.180
Guldstandard, 1g/l Au i 2% saltsyre AVS TITRINORM standard til AAS
Standarder
Kationiske standarder
Gold Standards
- Traceable to SRM from NIST
- Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
- Supplied with certificate of analysis, real value ±0,3%