Borstandard, 1g/l B i vand AVS TITRINORM standard til AAS
Leverandør: VWR Chemicals
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
86663.260EA
212.1
DKK
86663.260
86663.180
Borstandard, 1g/l B i vand AVS TITRINORM standard til AAS
Standarder
Kationiske standarder
Boron Standards
- Traceable to SRM from NIST
- Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
- Supplied with certificate of analysis, real value ±0,3%