Dodecyl β-D-glucopyranosid, white powder

Leverandør: MP Biomedicals
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Dodecyl b-D-glucopyranoside, Dodecyl glucoside, n-Dodecyl β-D-glucopyranoside

0215248483 0215248480 0215248491
ICNA0215248483EA 0 DKK
ICNA0215248483 ICNA0215248480 ICNA0215248491
Dodecyl β-D-glucopyranosid, white powder
Dodecyl β-D-glucopyranosid

Dodecyl β-D-glucopyranoside is a long chain alkyl glycopyranoside and a classical nonionic amphiphile surfactant.


N-Dodecyl β-D-glucopyranoside is a classical nonionic amphiphile surfactant, which used in colloid research and micelle development. It may be used as a reference compound in long-chain alkyl glucoside separation and analysis procedures.


Non-ionic saccharide detergent used for the solubilization of membrane-bound proteins, such as G-Protein-Coupled Receptors, in native state


Soluble in methanol (200 mg/4 ml).

Formel: C₁₈H₃₆O₆
Molekylvægt: 348.48 g/mol
Opbevaringstemperatur: Fryser
MDL Number: MFCD00063298
CAS-nummer: 59122-55-3
EINECS: 261-614-4

Bestil nu

Specifikationer Testresultater

Identity Test Passes
FTIR Conforms to Reference


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR