Diisopropylcarbodiimid 99%
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
115210250
115211000
115215000
ACRO115210250EA
964
DKK
ACRO115210250
ACRO115211000
ACRO115215000
Diisopropylcarbodiimid 99%
Diisopropylcarbodiimid
Formel:
C₇H₁₄N₂ Kogepunkt: 145…148 °C (760 mmHg) Massefylde: 0,81 g/cm³ Flammepunkt: 33 °C Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00065689 CAS-nummer: 693-13-0 EINECS: 211-743-7 UN nummer: 3384 ADR: 6.1, |
Specifikationer Testresultater
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4320 to 1.4350 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...