4-Amino-3-hydroxy-1-naphthalensulfonsyre 90+% ACS
Leverandør: Thermo Fisher Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
1-Amino-2-naphthol-4-sulphonic acid, 4-Amino-3-hydroxy-α-naphthalenesulphonic acid, 4-Amino-3-hydroxynaphthalene-1-sulphonic acid, 1-Amino-2-hydroxy-4-naphthalenesulphonic acid
400850250
400851000
ACRO400850250EA
918
DKK
ACRO400850250
ACRO400851000
4-Amino-3-hydroxy-1-naphthalensulfonsyre 90+% ACS
4-Amino-3-hydroxy-1-naphthalensulfonsyre
Formel:
C₁₀H₉NO₄S Kogepunkt: 466 °C (1013 hPa) Smeltepunkt: 295 °C (dec.) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00004019 CAS-nummer: 116-63-2 EINECS: 204-147-3 Merck Index: 13,00453 |
Specifikationer Testresultater
Appearance | White to light brown-pink to light purple Powder |
Infrared spectrum | Conforms |
Solubility | (in sodium carbonate) Passes test |
Residue after ignition | ≤0.1 % |
Titration with NaOH | ≥90.0 % |
Sensitivity | (to phosphate) Passes test |
Sulfate (SO4) | ≤0.2 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...