3-Amino-2-naphthoic acid 85% tech.
Leverandør: Thermo Fisher Scientific
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
104140050 104140250
ACRO104140050EA 846 DKK
ACRO104140050 ACRO104140250
3-Amino-2-naphthoic acid 85% tech.
3-Amino-2-naphthoic acid
Formel: C₁₁H₉NO₂
Kogepunkt: 383 °C (1013 hPa)
Smeltepunkt: 210 °C (dec.)
MDL Number: MFCD00004115
CAS-nummer: 5959-52-4
EINECS: 227-726-2
Merck Index: 13,00451
Bestil nu
Specifikationer Testresultater
Appearance Yellow to green Powder
Infrared spectrum Conforms
HPLC ≥80 %
Titration with HClO4 ≥80 %