Tetranitro blue tetrazolium

Leverandør: Serva
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: TNBT, 3,3'-(3,3'-Dimethoxy[1,1'-biphenyl]-4,4'-diyl)bis[2,5-bis(p-nitrophenyl)-2H-tetrazolium] dichloride

35935.02 35935.03
SERA35935.02EA 0 DKK
SERA35935.02 SERA35935.03
Tetranitro blue tetrazolium
Tetranitro blue tetrazolium
Formel: C₄₀H₂₈Cl₂N₁₂O₁₀
Smeltepunkt: 165 °C
MDL Number: MFCD00036338
CAS-nummer: 1184-43-6
EINECS: 214-665-1

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR