PTFE Tubings
Leverandør: MICROSOLV
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
49210-07-C 49210-20-C
MICS49210-07-CEA 730 DKK
MICS49210-07-C MICS49210-20-C
PTFE Tubings
Slanger Overførselsslange
PTFE (Polytetrafluoroethylene) commonly called Teflon™ is a lowest cost plastic or polymeric tubing and is widely used in all low-pressure laboratory applications.
  • PTFE tubing does contain micro-pores in the tubing wall allowing a quick air exchange
  • All MicroSolv PTFE tubing is extruded from virgin PTFE
  • No additives to the original PTFE material, except for colour particles in the solid colored tubing
  • Temperature limits are +260 and –240 °C
  • Low density, coherent fibrous structure
It is extruded from a fine fluoroplastic resin (641XTX) that has non-aging characteristics, exceptional dielectric properties which is stable with frequency and temperature, a low coefficient of friction and negligible moisture absorption.