N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimid, HCl for synthesis
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Advarsel
Synonymer: N'-(Ethylcarbonimidoyl)-N,N-dimethylpropane-1,3-diamine monohydrochloride, N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide monohydrochloride, 1-Ethyl-3-(3-dimethylaminopropyl)carbodiimide hydrochloride, EDC hydrochloride, EDC, EDAC HCl, EDAC hydrochloride, 1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide hydrochloride, WSC HCl, EDC HCl
M-Clarity™ quality level = MQ200
8.00907.9999 8.00907.0001 8.00907.0005 8.00907.0025
8.00907.9999EA 0 DKK
8.00907.9999 8.00907.0001 MERC8.00907.0005 MERC8.00907.0025
N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimid, HCl for synthesis
N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimid, HCl
Formel: C₈H₁₇N₃·HCl
Molekylvægt: 191.7 g/mol
Smeltepunkt: 112…113 °C
Opbevaringstemperatur: Fryser
MDL Number: MFCD00012503
CAS-nummer: 25952-53-8
EINECS: 247-361-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=25d8bf2864bcf21ffe6dcca6f22388d1fe427e22&vid=9080dd6b66a1622ef32a2da2138f85e44cc881b8&prid=web&clid=SAPDAM