Safety Shoes, Low ESD S3, MONSTER
Leverandør: U-Power
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
RL20366/38 RL20366/39 RL20366/40 RL20366/41 RL20366/42 RL20366/43 RL20366/44 RL20366/45 RL20366/46 RL20366/47 RL20366/48
UPOWRL20366/38EA 1330 DKK
UPOWRL20366/38 UPOWRL20366/39 UPOWRL20366/40 UPOWRL20366/41 UPOWRL20366/42 UPOWRL20366/43 UPOWRL20366/44 UPOWRL20366/45 UPOWRL20366/46 UPOWRL20366/47 UPOWRL20366/48
Safety Shoes, Low ESD S3, MONSTER
Sko og støvler
RedLion safety shoes, with air toe aluminium and flex PLUS anti-perforation system, designed to guarantee a prolonged well-being of the foot.
  • Lightweight
  • Comfortable
  • Removable template: Memory
  • High breathability and performance fit with BOA® fit system
  • Padded nylon bellows to ensure long lasting confort
These work shoes are able to guarantee comfort and lightness thanks to the sole with the innovative Infinergy® system and to the expanded PU midsole. The multi-functional heel and the compact PU tread provide grip, security and stability of posture.
Upper: New safety PUTEK® PLUS with very high resistance to abrasion, waterproof and breathable
Toe cap: AirToe® Aluminum with breathable membrane
Anti-perforation: Save and Flex PLUS®, non-metal anti-perforation insole
Midsole: Soft expanded PU foam and Infinergy®
Sole: PU, non-slip, anti-abrasion, oil resistant, antistatic
Fit: Natural comfort 11 mondopoint
Lining: Revolutionary ventilated WingTex® lining for maximum breathability. The special microcells have great absorbent and desorbent functions
Certificeringer: EN ISO 20345:2011.