AlphaTec® 53-003 Chemical Resistant Gloves
Leverandør: Ansell
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
53-003/7 53-003/8 53-003/9 53-003/10 53-003/11
112-6658EA 462 DKK
112-6658 112-6659 112-6660 112-6657 ANSE53-003/11
AlphaTec® 53-003 Chemical Resistant Gloves
Handsker
Nylon supported neoprene gloves that provide broad chemical protection with enhanced comfort and tactility.
  • MICROCHEM™ chemical barrier technology: Unique multi-layer rubber polymer and plastic laminate technology for extremely broad chemical protection
  • Multi-hazard protection: AlphaTec 53-003 provides next generational chemical protection whilst delivering secondary hazard protection, like mechanical hazards and heat hazards
  • Comfort and performance: Unique manufacturing process and nylon liner support to enhance comfort during use
Extreme Chemical protection: AlphaTec® 53-003 gloves achieving 17 out of 18 chemicals on EN 374:2016 with 16 being at class 6 >480 minutes and 16 out of 18> class 2 against ASTM F739 with a 10x lower limit to pass.
Elevated comfort and dexterity: Unique manufacturing process for optimised fit with raised diamond grip for secure performance in a wide range of scenarios.
Improved durability: They also possess EN 388-compliant abrasion resistance for better mechanical protection.
Specialized protection: AlphaTec® 53-003 gloves satisfy EN 407 contact heat level 1 (contact heat approved up to 100 °C/212 °F) , and anti-static according to EN 1149-3 standards preventing workplace injuries.
Silicone free and raised diamond grip design.
Primary industries: Automotive, chemical, recycling and waste disposal, utilities.
Recommended For:
Assembly and inspection of components
Oil, fluids and filter change
Painting or cleaning tools or robots
Chemical handling
Transferring liquids and solids
Chemical leaks
Spill or leakage cleanup
Certificeringer: EN ISO 374-1:2016 Type A (ABCEFGHIJKLMNOPST), EN ISO 374-5 VIRUS, EN 421, EN 388:2016 (2121X), EN 407 (X1XXXX)