Y-320
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Y0019-25MG
Y0019-5MG
TCIAY0019-25MGEA
1700
DKK
TCIAY0019-25MG
TCIAY0019-5MG
Y-320
Y-320
Formel:
C₂₇H₂₉ClN₆O₂ |
MDL Number:
MFCD28099803 CAS-nummer: 288250-47-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...