(3R,4R,5R)-3,4-Bis(benzyloxy)-5-((benzyloxy)methyl)dihydrofuran-2(3H)-one

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

T3955-25G T3955-5G
TCIAT3955-25GEA 1760 DKK
TCIAT3955-25G TCIAT3955-5G
(3R,4R,5R)-3,4-Bis(benzyloxy)-5-((benzyloxy)methyl)dihydrofuran-2(3H)-one
(3R,4R,5R)-3,4-Bis(benzyloxy)-5-((benzyloxy)methyl)dihydrofuran-2(3H)-one
Formel: C₂₆H₂₆O₅
MDL Number: MFCD08703966
CAS-nummer: 55094-52-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR