[6-(Trifluoromethyl)-3-pyridinyl]methanol

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

T3927-1G T3927-250MG
TCIAT3927-1GEA 903 DKK
TCIAT3927-1G TCIAT3927-250MG
[6-(Trifluoromethyl)-3-pyridinyl]methanol
[6-(Trifluoromethyl)-3-pyridinyl]methanol
Formel: C₇H₆F₃NO
MDL Number: MFCD01862648
CAS-nummer: 386704-04-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR