4'-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-[1,1'-biphenyl]-4-carbonitrile

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

T3912-1G T3912-5G
TCIAT3912-1GEA 501 DKK
TCIAT3912-1G TCIAT3912-5G
4'-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-[1,1'-biphenyl]-4-carbonitrile
4'-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-[1,1'-biphenyl]-4-carbonitrile
Formel: C₁₉H₂₀BNO₂
MDL Number: MFCD17015781
CAS-nummer: 406482-72-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR