Octaethyl (benzene-1,2,4,5-tetrayltetrakis(methylene))tetrakis(phosphonate)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: Octaethyl [benzene-1,2,4,5-tetrayltetrakis(methylene)]tetrakis (phosphonate)

T3904-1G T3904-5G
TCIAT3904-1GEA 411 DKK
TCIAT3904-1G TCIAT3904-5G
Octaethyl (benzene-1,2,4,5-tetrayltetrakis(methylene))tetrakis(phosphonate)
Octaethyl (benzene-1,2,4,5-tetrayltetrakis(methylene))tetrakis(phosphonate)
Formel: C₂₆H₅₀O₁₂P₄
Molekylvægt: 678.56 g/mol
CAS-nummer: 136455-49-7

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR