1,2,4,5-Tetrakis(bromomethyl)benzene

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

T3901-1G T3901-5G
TCIAT3901-1GEA 329 DKK
TCIAT3901-1G TCIAT3901-5G
1,2,4,5-Tetrakis(bromomethyl)benzene
1,2,4,5-Tetrakis(bromomethyl)benzene
Formel: C₁₀H₁₀Br₄
MDL Number: MFCD00009664
CAS-nummer: 15442-91-8
UN nummer: 3261

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR