2,2'-Thenil

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

T2901-1G T2901-5G
TCIAT2901-1GEA 329 DKK
TCIAT2901-1G TCIAT2901-5G
2,2'-Thenil
2,2'-Thenil
Formel: C₁₀H₆O₂S₂
MDL Number: MFCD00173678
CAS-nummer: 7333-07-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR