Ethyl acetopyruvate

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

R0165-25G R0165-5G
TCIAR0165-25GEA 2140 DKK
TCIAR0165-25G TCIAR0165-5G
Ethyl acetopyruvate
Ethyl acetopyruvate
Formel: C₇H₁₀O₄
MDL Number: MFCD00009124
CAS-nummer: 615-79-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR