Potassium-2,3,3-trimethyl-3H-indole-5-sulphonate
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
P2824-1G
P2824-5G
TCIAP2824-1GEA
542
DKK
TCIAP2824-1G
TCIAP2824-5G
Potassium-2,3,3-trimethyl-3H-indole-5-sulphonate
Potassium-2,3,3-trimethyl-3H-indole-5-sulphonate
Formel:
C₁₁H₁₂KNO₃S |
MDL Number:
MFCD07369684 CAS-nummer: 184351-56-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...