1-Phenyl-1H-imidazole trifluoromethanesulphonate

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 1-Phenyl-1H-imidazol-3-ium trifluoromethanesulphonate

P2822-25G P2822-5G
TCIAP2822-25GEA 3000 DKK
TCIAP2822-25G TCIAP2822-5G
1-Phenyl-1H-imidazole trifluoromethanesulphonate
1-Phenyl-1H-imidazole trifluoromethanesulphonate
Formel: C₁₀H₉F₃N₂O₃S
Molekylvægt: 294.250 g/mol
MDL Number: MFCD32206610
CAS-nummer: 361447-81-6

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR