3-(3-Pyridyl)-D-alanine dihydrochloride

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

P2805-1G P2805-250MG
TCIAP2805-1GEA 2000 DKK
TCIAP2805-1G TCIAP2805-250MG
3-(3-Pyridyl)-D-alanine dihydrochloride
3-(3-Pyridyl)-D-alanine dihydrochloride
Formel: C₈H₁₀N₂O₂·2ClH
MDL Number: MFCD00191179
CAS-nummer: 93960-21-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR