4-(Benzyloxy)benzenesulphonyl chloride

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

P2803-1G P2803-5G
TCIAP2803-1GEA 452 DKK
TCIAP2803-1G TCIAP2803-5G
4-(Benzyloxy)benzenesulphonyl chloride
4-(Benzyloxy)benzenesulphonyl chloride
Formel: C₁₃H₁₁ClO₃S
MDL Number: MFCD04035557
CAS-nummer: 87001-32-9
UN nummer: 3261

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR