(R)-4-Chloro-N-(quinuclidin-3-yl)benzamide
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
PNU-282987 free base
P2774-10MG
P2774-50MG
TCIAP2774-10MGEA
591
DKK
TCIAP2774-10MG
TCIAP2774-50MG
(R)-4-Chloro-N-(quinuclidin-3-yl)benzamide
(R)-4-Chloro-N-(quinuclidin-3-yl)benzamide
Formel:
C₁₄H₁₇ClN₂O |
MDL Number:
MFCD08703110 CAS-nummer: 711085-63-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...