Potassium (4-acetylphenyl)trifluoroborate

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

P2652-1G P2652-5G
TCIAP2652-1GEA 370 DKK
TCIAP2652-1G TCIAP2652-5G
Potassium (4-acetylphenyl)trifluoroborate
Potassium (4-acetylphenyl)trifluoroborate
Formel: C₈H₇BF₃KO
MDL Number: MFCD07784376
CAS-nummer: 252726-24-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR