Kalium(4-Bromphenyl)trifluorborat

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: (4-Bromophenyl)trifluoroboric acid potassium salt, Potassium (4-bromophenyl)trifluoroborate, Potassium 4-bromophenyltrifluoroborate

P2647-1G P2647-5G
TCIAP2647-1GEA 247 DKK
TCIAP2647-1G TCIAP2647-5G
Kalium(4-Bromphenyl)trifluorborat
Kalium(4-Bromphenyl)trifluorborat
Formel: C₆H₄BBrF₃K
MDL Number: MFCD02093978
CAS-nummer: 374564-35-9

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR