Picolinic acid N-oxide

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

P2516-25G P2516-5G
TCIAP2516-25GEA 2290 DKK
TCIAP2516-25G TCIAP2516-5G
Picolinic acid N-oxide
Picolinic acid N-oxide
Formel: C₆H₅NO₃
Molekylvægt: 139.1088 g/mol
MDL Number: MFCD00006198
CAS-nummer: 824-40-8

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR