(2-Aminoethoxy)benzen

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

P1663-25G P1663-5G
TCIAP1663-25GEA 1030 DKK
TCIAP1663-25G TCIAP1663-5G
(2-Aminoethoxy)benzen
(2-Aminoethoxy)benzen
Formel: C₆H₅OCH₂CH₂NH₂
MDL Number: MFCD00052975
CAS-nummer: 1758-46-9
UN nummer: 2735

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR