Oleuropein
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Methyl (2S,4S,E)-4-(2-(3,4-dihydroxyphenethoxy)-2-oxoethyl)-3-ethylidene-2-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-3,4-dihydro-2H-pyran-5-carboxylate
O0420-100MG
O0420-25MG
TCIAO0420-100MGEA
1490
DKK
TCIAO0420-100MG
TCIAO0420-25MG
Oleuropein
Oleuropein
Formel:
C₂₅H₃₂O₁₃ Molekylvægt: 540.51 g/mol |
MDL Number:
MFCD16879008 CAS-nummer: 32619-42-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...