Nickel(II) bromide 2-methoxyethyl ether complex

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

N1049-10G N1049-1G
TCIAN1049-10GEA 1060 DKK
TCIAN1049-10G TCIAN1049-1G
Nickel(II) bromide 2-methoxyethyl ether complex
Nickel(II) bromide 2-methoxyethyl ether complex
Formel: C₆H₁₄Br₂NiO₃
Molekylvægt: 352.6700 g/mol
MDL Number: MFCD00274339
CAS-nummer: 312696-09-6
UN nummer: 1325

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR