2-Methoxy-5-nitropyrimidine

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

M3080-1G M3080-5G
TCIAM3080-1GEA 550 DKK
TCIAM3080-1G TCIAM3080-5G
2-Methoxy-5-nitropyrimidine
2-Methoxy-5-nitropyrimidine
Formel: C₅H₅N₃O₃
MDL Number: MFCD00474639
CAS-nummer: 14001-69-5

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR