2,3'-Bipyridine
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
D1772-1G
D1772-5G
TCIAD1772-1GEA
353
DKK
TCIAD1772-1G
TCIAD1772-5G
2,3'-Bipyridine
2,3'-Bipyridine
Formel:
C₁₀H₈N₂ |
MDL Number:
MFCD00006215 CAS-nummer: 581-50-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...