Benzyl (6-hydroxyhexyl)carbamate

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 6-(N-Benzyloxycarbonylamino)-1-hexanol

B6075-1G B6075-5G
TCIAB6075-1GEA 345 DKK
TCIAB6075-1G TCIAB6075-5G
Benzyl (6-hydroxyhexyl)carbamate
Benzyl (6-hydroxyhexyl)carbamate
Formel: C₁₄H₂₁NO₃
Molekylvægt: 251.32 g/mol
MDL Number: MFCD00859036
CAS-nummer: 17996-12-2

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR